Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-11
Entrez Gene ID
175516
Gene Name
GLoBin related
Gene Aliases
CELE_C36E8.2
Chromosome Location
Chr.III: 3989298 - 3992189 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065401.5 NP_497802.2 3 - 4 567 95

View Full Product Details