Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
his-69
Entrez Gene ID
184005
Gene Name
Putative histone H3.3-like type 3
Gene Aliases
CELE_E03A3.3
Chromosome Location
Chr.III: 4058360 - 4058743 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065410.3 NP_497811.1 1 - 1 56 57

View Full Product Details