Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wht-3
Entrez Gene ID
175528
Gene Name
protein-coding
Gene Aliases
CELE_C16C10.12
Chromosome Location
Chr.III: 4147425 - 4151598 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065424.3 NP_497825.2 3 - 4 289 93

View Full Product Details