Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnj-16
Entrez Gene ID
175540
Gene Name
protein-coding
Gene Aliases
CELE_R74.4
Chromosome Location
Chr.III: 4191141 - 4193429 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267961.2 NP_001254890.1 2 - 3 231 77
NM_001267962.2 NP_001254891.1 1 - 2 94 77

View Full Product Details