Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mpk-1
Entrez Gene ID
175545
Gene Name
protein-coding
Gene Aliases
CELE_F43C1.2
Chromosome Location
Chr.III: 4216764 - 4228108 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027413.4 NP_001022584.1 3 - 4 329 87
NM_001381825.1 NP_001366708.1 3 - 4 343 87
GenBank mRNA U27124.1 - 2 - 3 125 87

View Full Product Details