Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mpk-1
Entrez Gene ID
175545
Gene Name
protein-coding
Gene Aliases
CELE_F43C1.2
Chromosome Location
Chr.III: 4216764 - 4228108 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027413.4 NP_001022584.1 1 - 2 207 66
NM_001381825.1 NP_001366708.1 1 - 2 221 66

View Full Product Details