Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-20
Entrez Gene ID
175546
Gene Name
Nuclear hormone receptor family member nhr-20
Gene Aliases
CELE_F43C1.4
Chromosome Location
Chr.III: 4230434 - 4233219 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065448.6 NP_497849.1 3 - 4 482 71

View Full Product Details