Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
evl-14
Entrez Gene ID
175555
Gene Name
Uncharacterized protein
Gene Aliases
CELE_H38K22.1
Chromosome Location
Chr.III: 4303096 - 4313168 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065464.8 NP_497865.2 15 - 16 2929 142

View Full Product Details