Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mnp-1
Entrez Gene ID
175564
Gene Name
Matrix non-peptidase homolog 1
Gene Aliases
CELE_B0285.7
Chromosome Location
Chr.III: 4356594 - 4364979 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065477.6 NP_497878.1 2 - 3 794 88

View Full Product Details