Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cku-80
Entrez Gene ID
175578
Gene Name
protein-coding
Gene Aliases
CELE_R07E5.8
Chromosome Location
Chr.III: 4419848 - 4423375 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322514.2 NP_001309438.1 8 - 9 1366 136
NM_001322520.3 NP_001309599.1 1 - 2 215 136

View Full Product Details