Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ndg-4
Entrez Gene ID
175582
Gene Name
protein-coding
Gene Aliases
CELE_F56F3.2
Chromosome Location
Chr.III: 4462833 - 4467377 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027447.6 NP_001022618.1 2 - 3 344 97
NM_001027448.4 NP_001022619.1 2 - 3 344 97
GenBank mRNA AF173373.1 - 2 - 3 342 97

View Full Product Details