Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-13
Entrez Gene ID
191839
Gene Name
Serpentine receptor class gamma-13
Gene Aliases
CELE_T23F11.5
Chromosome Location
Chr.III: 4672249 - 4674207 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065552.2 NP_497953.1 2 - 3 449 146

View Full Product Details