Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnj-18
Entrez Gene ID
175616
Gene Name
J domain-containing protein
Gene Aliases
CELE_T04A8.9
Chromosome Location
Chr.III: 4698119 - 4699454 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065561.6 NP_497962.1 1 - 2 216 116

View Full Product Details