Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rps-0
- Entrez Gene ID
-
175628
- Gene Name
- 40S ribosomal protein SA
- Gene Aliases
-
CELE_B0393.1
- Chromosome Location
-
Chr.III: 4752011 - 4754051 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065577.8 |
NP_497978.1 |
2 - 3 |
398 |
78 |
- Gene Symbol
- B0393.8
- Entrez Gene ID
-
181959
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_B0393.8
- Chromosome Location
-
Chr.III: 4751878 - 4754057 on Build WBcel235
View Full Product Details