Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mel-28
Entrez Gene ID
175634
Gene Name
Protein mel-28
Gene Aliases
CELE_C38D4.3
Chromosome Location
Chr.III: 4786080 - 4792391 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065586.7 NP_497987.2 16 - 17 5176 81

View Full Product Details