Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rom-1
Entrez Gene ID
184989
Gene Name
Rhomboid-related protein 1
Gene Aliases
CELE_F26F4.3
Chromosome Location
Chr.III: 4907196 - 4908993 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065628.4 NP_498029.2 1 - 2 229 74

View Full Product Details