Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cee-1
Entrez Gene ID
175658
Gene Name
Conserved Edge Expressed protein
Gene Aliases
CELE_F26F4.1
Chromosome Location
Chr.III: 4912207 - 4914196 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065633.10 NP_498034.1 4 - 5 890 116

View Full Product Details