Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mmab-1
Entrez Gene ID
175661
Gene Name
Corrinoid adenosyltransferase
Gene Aliases
CELE_C26E6.11
Chromosome Location
Chr.III: 4918330 - 4922114 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001384085.1 3 - 4 450 108

View Full Product Details