Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
flh-2
Entrez Gene ID
175670
Gene Name
FLYWCH-type domain-containing protein
Gene Aliases
CELE_C26E6.2
Chromosome Location
Chr.III: 4952124 - 4955171 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065648.6 NP_498049.2 4 - 5 1204 67

View Full Product Details