Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nca-2
Entrez Gene ID
175674
Gene Name
protein-coding
Gene Aliases
CELE_C27F2.2
Chromosome Location
Chr.III: 4965571 - 4979738 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136363.4 NP_001129835.1 11 - 12 2678 92
NM_065653.6 NP_498054.2 12 - 13 2834 92
GenBank mRNA AY555272.2 - 12 - 13 2834 92
DQ917241.1 - 11 - 12 2716 92

View Full Product Details