Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
larp-1
Entrez Gene ID
175680
Gene Name
protein-coding
Gene Aliases
CELE_R144.7
Chromosome Location
Chr.III: 5008925 - 5017650 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047403.5 NP_001040868.3 5 - 6 995 88

View Full Product Details