Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
klp-6
Entrez Gene ID
175683
Gene Name
Kinesin-like protein
Gene Aliases
CELE_R144.1
Chromosome Location
Chr.III: 5030929 - 5036583 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065668.6 NP_498069.1 9 - 10 1937 115
GenBank mRNA AB023497.1 - 9 - 10 1937 115

View Full Product Details