Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
alh-1
Entrez Gene ID
175691
Gene Name
protein-coding
Gene Aliases
CELE_F54D8.3
Chromosome Location
Chr.III: 5077763 - 5080674 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065680.7 NP_498081.2 2 - 3 221 74

View Full Product Details