The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322586.2 | NP_001309492.1 | - | 974 | 102 |
NM_001322587.1 | NP_001309597.1 | 3 - 4 | 492 | 102 |