Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cnd-1
Entrez Gene ID
183212
Gene Name
Neurogenic differentiation factor 1
Gene Aliases
CELE_C34E10.7
Chromosome Location
Chr.III: 5240584 - 5244640 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065714.6 NP_498115.1 2 - 3 564 148

View Full Product Details