Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gop-1
Entrez Gene ID
175721
Gene Name
Protein CLEC16A homolog
Gene Aliases
CELE_C34E10.3
Chromosome Location
Chr.III: 5254044 - 5258942 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065716.6 NP_498117.3 15 - 16 2446 103

View Full Product Details