Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nekl-4
Entrez Gene ID
175760
Gene Name
Serine/threonine-protein kinase D1044.8
Gene Aliases
CELE_D1044.8
Chromosome Location
Chr.III: 5512912 - 5517523 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065777.7 NP_498178.3 1 - 2 109 131

View Full Product Details