Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
uev-2
Entrez Gene ID
186377
Gene Name
UBIQUITIN_CONJUGAT_2 domain-containing protein
Gene Aliases
CELE_F56D2.4
Chromosome Location
Chr.III: 5583125 - 5584522 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065797.3 NP_498198.1 2 - 3 284 84

View Full Product Details