Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ckk-1
Entrez Gene ID
182278
Gene Name
protein-coding
Gene Aliases
CELE_C05H8.1
Chromosome Location
Chr.III: 5635479 - 5654564 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025981.3 NP_001021152.1 7 - 8 1399 88
NM_001025982.5 NP_001021153.1 10 - 11 1536 88
GenBank mRNA AB016838.4 - 7 - 8 1399 88

View Full Product Details