Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pst-2
Entrez Gene ID
175787
Gene Name
protein-coding
Gene Aliases
CELE_F54E7.1
Chromosome Location
Chr.III: 5680433 - 5682325 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047390.4 NP_001040855.1 4 - 5 568 141
NM_001047391.4 NP_001040856.1 1 - 2 152 141

View Full Product Details