Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
polq-1
Entrez Gene ID
175810
Gene Name
DNA polymerase theta
Gene Aliases
CELE_W03A3.2
Chromosome Location
Chr.III: 5791119 - 5803160 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379729.2 NP_001367994.1 17 - 18 4533 149

View Full Product Details