Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pxl-1
Entrez Gene ID
175831
Gene Name
protein-coding
Gene Aliases
CELE_C28H8.6
Chromosome Location
Chr.III: 5889721 - 5896017 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026014.4 NP_001021185.2 7 - 8 732 96
NM_001026015.3 NP_001021186.1 3 - 4 530 96
NM_001129205.3 NP_001122677.1 5 - 6 555 96
GenBank mRNA EU239658.1 - 7 - 8 727 96
EU239659.1 - 3 - 4 466 96
EU239660.1 - 5 - 6 544 96

View Full Product Details