Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npp-10
Entrez Gene ID
175852
Gene Name
protein-coding
Gene Aliases
CELE_ZK328.5
Chromosome Location
Chr.III: 6013447 - 6019562 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379737.1 NP_001367842.1 7 - 8 3440 112
NM_065909.5 NP_498310.3 7 - 8 3441 112
GenBank mRNA GU174496.1 - 7 - 8 3430 112

View Full Product Details