Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyk-3
Entrez Gene ID
175853
Gene Name
protein-coding
Gene Aliases
CELE_ZK328.1
Chromosome Location
Chr.III: 6020159 - 6024985 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027821.6 NP_001022992.1 7 - 8 1135 149
NM_065910.7 NP_498311.2 7 - 8 1129 149
GenBank mRNA AF469173.1 - 7 - 8 1082 149

View Full Product Details