Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbx-7
Entrez Gene ID
260030
Gene Name
protein-coding
Gene Aliases
CELE_ZK328.8
Chromosome Location
Chr.III: 6024958 - 6028532 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379738.1 NP_001368540.1 3 - 4 543 131
NM_001379739.1 NP_001368539.1 3 - 4 568 131

View Full Product Details