Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nfm-1
Entrez Gene ID
175870
Gene Name
protein-coding
Gene Aliases
CELE_F42A10.2
Chromosome Location
Chr.III: 6159453 - 6164589 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065935.5 NP_498336.1 10 - 11 2298 69

View Full Product Details