Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpn-2
Entrez Gene ID
175877
Gene Name
protein-coding
Gene Aliases
CELE_C23G10.4
Chromosome Location
Chr.III: 6198902 - 6202594 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065945.6 NP_498346.2 6 - 7 2641 82

View Full Product Details