Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-6
Entrez Gene ID
24104889
Gene Name
Serpentine receptor class gamma-6
Gene Aliases
CELE_T12A2.13
Chromosome Location
Chr.III: 6264460 - 6266011 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306704.1 NP_001293633.1 1 - 2 202 102

View Full Product Details