Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
thoc-2
Entrez Gene ID
175900
Gene Name
THO Complex (transcription factor/nuclear export) subunit
Gene Aliases
CELE_C16A3.8
Chromosome Location
Chr.III: 6368042 - 6373908 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065991.7 NP_498392.2 11 - 12 2859 103

View Full Product Details