Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vps-16
Entrez Gene ID
175913
Gene Name
Vacuolar protein sorting-associated protein 16 homolog
Gene Aliases
CELE_C05D11.2
Chromosome Location
Chr.III: 6433239 - 6436502 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066010.6 NP_498411.1 2 - 3 186 79

View Full Product Details