Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wht-6
Entrez Gene ID
188911
Gene Name
ABC transporter domain-containing protein
Gene Aliases
CELE_T26A5.1
Chromosome Location
Chr.III: 6471714 - 6475246 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066024.3 NP_498425.2 3 - 4 803 94

View Full Product Details