Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acr-5
Entrez Gene ID
191596
Gene Name
Acetylcholine receptor subunit alpha-type acr-5
Gene Aliases
CELE_K03F8.2
Chromosome Location
Chr.III: 6509445 - 6514773 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379767.1 NP_001367477.1 8 - 9 1171 104

View Full Product Details