Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oig-1
Entrez Gene ID
182450
Gene Name
Immunoglobulin domain-containing protein oig-1
Gene Aliases
CELE_C09E7.3
Chromosome Location
Chr.III: 6524051 - 6525660 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066038.2 NP_498439.1 2 - 3 311 140

View Full Product Details