Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pdfr-1
Entrez Gene ID
175942
Gene Name
protein-coding
Gene Aliases
CELE_C13B9.4
Chromosome Location
Chr.III: 6632837 - 6652083 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025999.4 NP_001021170.1 15 - 16 2262 76
NM_001026000.4 NP_001021171.1 15 - 16 2232 76
NM_001372610.2 NP_001360187.1 13 - 14 1589 76
NM_001382898.1 13 - 14 1619 76
GenBank mRNA AY314776.1 - 15 - 16 2232 76
AY314777.1 - 15 - 16 2262 76
AY314778.1 - 15 - 16 2262 76
EF141316.1 - 12 - 13 1540 76
EF141317.1 - 12 - 13 1510 76
EF141318.1 - 12 - 13 1540 76

View Full Product Details