Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lim-8
Entrez Gene ID
175952
Gene Name
protein-coding
Gene Aliases
CELE_F28F5.3
Chromosome Location
Chr.III: 6802788 - 6816141 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066083.7 NP_498484.1 1 - 2 429 90

View Full Product Details