Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sma-3
Entrez Gene ID
175955
Gene Name
Dwarfin sma-3
Gene Aliases
CELE_R13F6.9
Chromosome Location
Chr.III: 6860939 - 6863968 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066092.4 NP_498493.1 3 - 4 306 96
GenBank mRNA U34902.1 - 3 - 4 307 96

View Full Product Details