Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
brd-1
Entrez Gene ID
175959
Gene Name
BRCA1-associated RING domain protein 1
Gene Aliases
CELE_K04C2.4
Chromosome Location
Chr.III: 6885202 - 6889504 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066097.8 NP_498498.3 4 - 5 478 103

View Full Product Details