Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rfc-4
Entrez Gene ID
175976
Gene Name
Replication factor C subunit 4
Gene Aliases
CELE_F31E3.3
Chromosome Location
Chr.III: 6972210 - 6973717 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066120.7 NP_498521.1 4 - 5 352 63

View Full Product Details