Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-37
Entrez Gene ID
175986
Gene Name
Uncharacterized protein
Gene Aliases
CELE_ZK418.4
Chromosome Location
Chr.III: 7070800 - 7072552 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066142.5 NP_498543.1 4 - 5 715 107

View Full Product Details