Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpg-2
Entrez Gene ID
175991
Gene Name
Chondroitin proteoglycan-2
Gene Aliases
CELE_B0280.5
Chromosome Location
Chr.III: 7102086 - 7104121 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066150.13 NP_498551.3 5 - 6 1422 68
GenBank mRNA DQ340624.1 - 4 - 5 1370 68

View Full Product Details